accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3591-42-2

H3CClCl

CAS RN 3591-42-2

IUPAC Name: (2,2-dichloro-1-methylcyclopropyl)benzene
Synonyms: (2,2-dichloro-1-methylcyclopropyl)benzene
Molecular Weight (g/mol): 201.09
Molecular Formula: C10H10Cl2
InChi Key: NXYPRVQXSWVEOB-UHFFFAOYNA-N
SMILES: CC1(CC1(Cl)Cl)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)