missing translation for 'onlineSavingsMsg'
Learn More

(2,2-Dichloro-1-methylcyclopropyl)benzene, 95%, Thermo Scientific™

Product Code. 11427256
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427256 25 g 25g
11417256 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11427256 Supplier Thermo Scientific Alfa Aesar Supplier No. B25571.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3591-42-2
Molecular Formula C10H10Cl2
Molecular Weight (g/mol) 201.09
MDL Number MFCD00012260
InChI Key NXYPRVQXSWVEOB-UHFFFAOYSA-N
Synonym 2,2-dichloro-1-methylcyclopropyl benzene, benzene, 2,2-dichloro-1-methylcyclopropyl, 1,1-dichloro-2-methyl-2-phenylcyclopropane, 2,2-dichloro-1-methyl cyclopropyl benzene, 2,2-dichloro-1-methyl-cyclopropyl benzene, acmc-20aok2, 1-phenyl-1-methyl-2,2-dichlorocyclopropane, 1-methyl-2,2-dichlorocyclopropylbenzene, 2,2-dichloro-1-methyl-cyclopropyl-benzene, 2,2-dichloro-1-methyl-1-phenyl-cyclopropane
PubChem CID 107129
IUPAC Name (2,2-dichloro-1-methylcyclopropyl)benzene
SMILES CC1(CC1(Cl)Cl)C2=CC=CC=C2

Specifications

Boiling Point 70°C to 72°C (0.8 mmHg)
Refractive Index 1.541
Quantity 25 g
Formula Weight 201.09
Percent Purity 95%
Chemical Name or Material (2,2-Dichloro-1-methylcyclopropyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.