accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 36282-26-5

IUPAC Name: 2-bromo-4-fluorobenzonitrile
Synonyms: 2-bromo-4-fluorobenzonitrile
Molecular Weight (g/mol): 200.01
Molecular Formula: C7H3BrFN
InChi Key: MNNDREXLRLDWEY-UHFFFAOYSA-N
SMILES: FC1=CC=C(C#N)C(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results