missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorobenzonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11394556
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394556 5 g 5g
11384556 1 g 1g
11304566 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394556 Supplier Thermo Scientific Alfa Aesar Supplier No. L20115.06
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36282-26-5 | Molecular Formula: C7H3BrFN | Molecular Weight: 200.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36282-26-5
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD00672924
InChI Key MNNDREXLRLDWEY-UHFFFAOYSA-N
Synonym 4-fluoro-2-bromobenzonitrile, 2-bromo-4-fluoro-benzonitrile, 2-bromo-4-fluorobenzenecarbonitrile, 2-cyano-5-fluorobromobenzene, benzonitrile, 2-bromo-4-fluoro, pubchem3217, acmc-1ajip, ksc490q6h, 2-bromo-4-fluoro benzonitrile, 2-bromo-4-fluorobenzonitrile
PubChem CID 118939
IUPAC Name 2-bromo-4-fluorobenzonitrile
SMILES FC1=CC=C(C#N)C(Br)=C1

Specifications

Melting Point 74°C to 78°C
Quantity 5 g
UN Number UN3439
Beilstein 1940597
Formula Weight 200.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-fluorobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.