accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOCH3

CAS RN 3648-21-3

IUPAC Name: 1,2-diheptyl benzene-1,2-dicarboxylate
Synonyms: DHPP di-n-heptylphthalate N-diheptyl phthalate
Molecular Weight (g/mol): 362.51
Molecular Formula: C22H34O4
InChi Key: JQCXWCOOWVGKMT-UHFFFAOYSA-N
SMILES: CCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCC
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)