accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 36546-50-6

(S)OHNOHOCH3CH3OH2O

CAS RN 36546-50-6

IUPAC Name: ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate
Synonyms: acetyl-L-tyrosine ethyl ester ethyl N-acetyl-L-tyrosinate
Molecular Weight (g/mol): 251.28
Molecular Formula: C13H17NO4
InChi Key: SKAWDTAMLOJQNK-LBPRGKRZSA-N
SMILES: CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)