accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COH

CAS RN 36653-82-4

IUPAC Name: hexadecan-1-ol
Synonyms: cetyl alcohol exxal 16 eutanol G16 normal primary hexadecyl alcohol lorol 24 epa pesticide chemical code 001508 fema number 2554 N-hexadecyl alcohol ceraphyl ica cetyl alcohol (substance)
Molecular Weight (g/mol): 242.45
Molecular Formula: C16H34O
InChi Key: BXWNKGSJHAJOGX-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCO
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Boiling Point 
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)