accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OIOI

CAS RN 36839-55-1

IUPAC Name: 1-iodo-2-[2-(2-iodoethoxy)ethoxy]ethane
Synonyms: 1-iodo-2-[2-(2-iodoethoxy)ethoxy]ethane
Molecular Weight (g/mol): 369.97
Molecular Formula: C6H12I2O2
InChi Key: BCAGFJXMCZSAHD-UHFFFAOYSA-N
SMILES: ICCOCCOCCI

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results