accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3H3CCH3SOONNNCH3CH3

CAS RN 36982-84-0

IUPAC Name: 2,4,6-tris(propan-2-yl)benzene-1-sulfonyl azide
Synonyms: 2,4,6-triisopropylbenzenesulfonyl azide
Molecular Weight (g/mol): 309.43
Molecular Formula: C15H23N3O2S
InChi Key: AEMWUHCKKDPRSK-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(=O)(=O)N=[N+]=[N-]
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)