missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzenesulfonyl Azide, TRC

Product Code. 30453154
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453154 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453154 Supplier Toronto Research Chemicals Supplier No. TRCT79548025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl Azide
CAS 36982-84-0
Molecular Formula C15H23N3O2S
Quantity 25 g
Synonym 2,4,6-Triisopropylphenylsulfonyl Azide, 2,4,6-Tris(isopropyl)benzenesulfonyl azide, NSC 646156, Trisyl Azide
InChI Formula InChI=1S/C15H23N3O2S/c1-9(2)12-7-13(10(3)4)15(14(8-12)11(5)6)21(19,20)18-17-16/h7-11H,1-6H3
SMILES O=[S](N=[N+]=[N-])(C(C(C(C)C)=CC(C(C)C)=C1)=C1C(C)C)=O
IUPAC Name 2,4,6-triisopropylbenzenesulfonyl azide
Molecular Weight (g/mol) 309.43
Formula Weight 309.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.