CAS RN 371753-20-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac cis-Loxoprofen Alcohol, TRC
CAS: 371753-20-7 Molecular Formula: C15H20O3 Molecular Weight (g/mol): 248.32 Synonym: rel-4-[[(1R,2R)-2-Hydroxycyclopentyl]methyl]-alpha-methylbenzeneacetic Acid; IUPAC Name: 2-[4-[[(1R,2R)-2-hydroxycyclopentyl]methyl]phenyl]propanoic acid SMILES: CC(C(=O)O)c1ccc(C[C@H]2CCC[C@H]2O)cc1