missing translation for 'onlineSavingsMsg'
Learn More

rac cis-Loxoprofen Alcohol, TRC

Product Code. 30179296
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
1mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179296 25 mg 25mg
30178928 1 mg 1mg
30178975 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179296 Supplier Toronto Research Chemicals Supplier No. TRCL47291025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371753-20-7
Molecular Formula C15H20O3
Molecular Weight (g/mol) 248.32
Synonym rel-4-[[(1R,2R)-2-Hydroxycyclopentyl]methyl]-alpha-methylbenzeneacetic Acid;
IUPAC Name 2-[4-[[(1R,2R)-2-hydroxycyclopentyl]methyl]phenyl]propanoic acid
SMILES CC(C(=O)O)c1ccc(C[C@H]2CCC[C@H]2O)cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H20O3/c1-10(15(17)18)12-7-5-11(6-8-12)9-13-3-2-4-14(13)16/h5-8,10,13-14,16H,2-4,9H2,1H3,(H,17,18)/t10?,13-,14-/m1/s1
Formula Weight 248.1412
Chemical Name or Material rac cis-Loxoprofen Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.