accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClRuClClRuCl

CAS RN 37366-09-9

IUPAC Name: bis(benzene); bis(dichlororuthenium)
Synonyms: bis(benzene); bis(dichlororuthenium)
Molecular Weight (g/mol): 500.17
Molecular Formula: C12H12Cl4Ru2
InChi Key: YGXMUPKIEHNBNQ-UHFFFAOYSA-J
SMILES: Cl[Ru]Cl.Cl[Ru]Cl.C1=CC=CC=C1.C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results