missing translation for 'onlineSavingsMsg'
Learn More

Benezeneruthenium(II) Chloride Dimer, TRC

Product Code. 30329485
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329485 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329485 Supplier Toronto Research Chemicals Supplier No. TRCB997115250MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benezeneruthenium(II) Chloride Dimer
CAS 37366-09-9
Molecular Formula C12H12Cl4Ru2
Quantity 250 mg
Synonym Bis(eta6-benzene)di-μ-chlorodichlorodiruthenium, (Benzene)dichlororuthenium DimerBenzeneruthenium Dichloride Dimer, Bis((eta6-benzene)chloro(μ-chloro)ruthenium), Bis((eta6-benzene)dichlororuthenium), Bis(benzene)di-μ-chlorodichlorodiruthenium, Bis(benzene)tetrachlorodiruthenium, Bis(eta6-benzene)tetrachlorodiruthenium, Bis(π-benzeneruthenium dichloride), Bis[(benzene)dichlororuthenium], Bis[dichloro[eta6-(benzene)]ruthenium], Dibenzenetetrachlorodiruthenium, μ-Dichloro-bis(benzenechlororuthenium(II))
InChI Formula InChI=1S/2C6H6.4ClH.2Ru/c2*1-2-4-6-5-3-1;;;;;;/h2*1-6H;4*1H;;/q;;;;;;2*+2/p-4
SMILES [Cl-][Ru+2]1[Cl-][Ru+2]([Cl-])[Cl-]1.[cH]2[cH][cH][cH][cH][cH]2.[cH]3[cH][cH][cH][cH][cH]3
IUPAC Name benzene;dichlororuthenium
Molecular Weight (g/mol) 500.18
Formula Weight 499.778
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.