CAS RN 374538-02-0
IUPAC Name:
5-(3-phenoxyphenyl)-2H-1,2,3,4-tetrazole
Synonyms:
5-(3-phenoxyphenyl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol):
238.25
Molecular Formula:
C13H10N4O
InChi Key:
TYNITQBHBADQAR-UHFFFAOYSA-N
SMILES:
N1N=NC(=N1)C1=CC(OC2=CC=CC=C2)=CC=C1
1
–
1
of
1
results
1
Alfa Aesar™ 5-(3-Phenoxyphenyl)-1H-tetrazole, 98%
CAS: 374538-02-0 Molecular Formula: C13H10N4O Molecular Weight (g/mol): 238.25 MDL Number: MFCD02093975 InChI Key: TYNITQBHBADQAR-UHFFFAOYSA-N Synonym: 5-3-phenoxyphenyl-1h-tetrazole,5-3-phenoxyphenyl-2h-tetrazole,5-3-phenoxyphenyl-1h-1,2,3,4-tetrazole,acmc-1ctsy,2h-tetrazole,5-3-phenoxyphenyl,3-1h-1,2,3,4-tetraazol-5-yl-1-phenoxybenzene,5-3-phenoxyphenyl-2h-1,2,3,4-tetrazole PubChem CID: 7010320 IUPAC Name: 5-(3-phenoxyphenyl)-2H-tetrazole SMILES: C1=CC=C(C=C1)OC2=CC=CC(=C2)C3=NNN=N3