missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-(3-Phenoxyphenyl)-1H-tetrazole, 98%

Product Code. 11384835
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384835 250 mg 250mg
11374835 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384835 Supplier Alfa Aesar™ Supplier No. L17955.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 374538-02-0
Molecular Formula C13H10N4O
Molecular Weight (g/mol) 238.25
MDL Number MFCD02093975
InChI Key TYNITQBHBADQAR-UHFFFAOYSA-N
Synonym 5-3-phenoxyphenyl-1h-tetrazole, 5-3-phenoxyphenyl-2h-tetrazole, 5-3-phenoxyphenyl-1h-1,2,3,4-tetrazole, acmc-1ctsy, 2h-tetrazole,5-3-phenoxyphenyl, 3-1h-1,2,3,4-tetraazol-5-yl-1-phenoxybenzene, 5-3-phenoxyphenyl-2h-1,2,3,4-tetrazole
PubChem CID 7010320
IUPAC Name 5-(3-phenoxyphenyl)-2H-tetrazole
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)C3=NNN=N3

Specifications

Melting Point 152°C to 154°C
Quantity 250 mg
Formula Weight 238.25
Percent Purity 98%
Chemical Name or Material 5-(3-Phenoxyphenyl)-1H-tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.