accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 37473-67-9

OOOOOOCHHCCHH3CCH3CH3NdCF3CF3CF3

CAS RN 37473-67-9

IUPAC Name: tris(5,5,5-trifluoro-4-hydroxypent-3-en-2-one) neodymium
Synonyms: tris(5,5,5-trifluoro-4-hydroxypent-3-en-2-one) neodymium
Molecular Weight (g/mol): 606.51
Molecular Formula: C15H15F9NdO6
InChi Key: ODGRTCBLUZXKII-UHFFFAOYSA-N
SMILES: [Nd].CC(=O)C=C(O)C(F)(F)F.CC(=O)C=C(O)C(F)(F)F.CC(=O)C=C(O)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results