accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOH3CNNHHCl

CAS RN 374897-99-1

IUPAC Name: 1-[(2,5-dimethoxyphenyl)methyl]piperazine hydrochloride
Synonyms: 1-[(2,5-dimethoxyphenyl)methyl]piperazine hydrochloride
Molecular Weight (g/mol): 272.77
Molecular Formula: C13H21ClN2O2
InChi Key: UWSVJUWWZFXWEZ-UHFFFAOYSA-N
SMILES: Cl.COC1=CC(CN2CCNCC2)=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results