accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3O

CAS RN 37592-72-6

IUPAC Name: 1-(4-hexylphenyl)ethan-1-one
Synonyms: 1-(4-hexylphenyl)ethanone
Molecular Weight (g/mol): 204.31
Molecular Formula: C14H20O
InChi Key: WWBVHJKFJZBRSO-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(C=C1)C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results