accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 37593-05-8

H3CCH3O

CAS RN 37593-05-8

IUPAC Name: 1-(4-nonylphenyl)ethan-1-one
Synonyms: 1-(4-nonylphenyl)ethanone
Molecular Weight (g/mol): 246.39
Molecular Formula: C17H26O
InChi Key: PWRSUUUQEQAMEV-UHFFFAOYSA-N
SMILES: CCCCCCCCCC1=CC=C(C=C1)C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results