accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3761-94-2

CH3HO

CAS RN 3761-94-2

IUPAC Name: 1-methylcycloheptan-1-ol
Synonyms: 1-methyl-cycloheptanol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: XFFKAYOHINCUNU-UHFFFAOYSA-N
SMILES: CC1(O)CCCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results