accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NBrOH

CAS RN 37669-64-0

IUPAC Name: (5-bromopyridin-3-yl)methanol
Synonyms: (5-bromopyridin-3-yl)methanol
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: WDVDHJLKXYCOFS-UHFFFAOYSA-N
SMILES: OCC1=CC(Br)=CN=C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results