accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COO(S)HN

CAS RN 37675-18-6

IUPAC Name: ethyl (3S)-piperidine-3-carboxylate
Synonyms: ethyl (3S)-piperidine-3-carboxylate
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: XIWBSOUNZWSFKU-ZETCQYMHSA-N
SMILES: CCOC(=O)[C@H]1CCCNC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results