accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFClO

CAS RN 384-67-8

IUPAC Name: 2-chloro-2,2-difluoro-1-phenylethan-1-one
Synonyms: 2-chloro-2,2-difluoro-1-phenylethanone
Molecular Weight (g/mol): 190.57
Molecular Formula: C8H5ClF2O
InChi Key: MNOONJNILVDLSW-UHFFFAOYSA-N
SMILES: FC(F)(Cl)C(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results