accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3O

CAS RN 38861-78-8

IUPAC Name: 1-[4-(2-methylpropyl)phenyl]ethan-1-one
Synonyms: 1-[4-(2-methylpropyl)phenyl]ethanone
Molecular Weight (g/mol): 176.26
Molecular Formula: C12H16O
InChi Key: KEAGRYYGYWZVPC-UHFFFAOYSA-N
SMILES: CC(C)CC1=CC=C(C=C1)C(C)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)