accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOH3CCH3CH3O

CAS RN 38923-57-8

IUPAC Name: methyl 2,2-dimethyl-3-oxobutanoate
Synonyms: methyl 2,2-dimethyl-3-oxobutanoate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: LVSDLZIEHYYLTC-UHFFFAOYSA-N
SMILES: COC(=O)C(C)(C)C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results