missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,2-dimethylacetoacetate, 99%

Product Code. 11393066
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393066 1 g 1g
11303076 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393066 Supplier Thermo Scientific Alfa Aesar Supplier No. L10917.03
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38923-57-8 | C7H12O3 | 144.17 g/mol

Pyrimidine was synthesis from methyl 2, 2-dimethylacetoacetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrimidine was synthesis from methyl 2, 2-dimethylacetoacetate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38923-57-8
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00026232
InChI Key LVSDLZIEHYYLTC-UHFFFAOYSA-N
Synonym methyl 2,2-dimethylacetoacetate, methyl2,2-dimethylacetoacetate, butanoic acid, 2,2-dimethyl-3-oxo-, methyl ester, methyl 2,2-dimethyl acetoacetate, 2,2-dimethylacetoacetic acid methyl ester, 2,2-dimethyl-3-oxo-butyric acid methyl ester, 2,2-dimethyl-3-oxobutyric acid, methyl ester, methyl 2,2-dimethyl-3-oxobutyrate, pubchem23356, acmc-20akj7
PubChem CID 96925
IUPAC Name methyl 2,2-dimethyl-3-oxobutanoate
SMILES COC(=O)C(C)(C)C(C)=O

Specifications

Density 0.999
Boiling Point 72°C to 74°C (15 mmHg)
Refractive Index 1.4205
Quantity 1 g
Beilstein 1753940
Solubility Information Insoluble in water.
Formula Weight 144.17
Percent Purity 99%
Chemical Name or Material Methyl 2,2-dimethylacetoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.