accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 38966-21-1

IUPAC Name: (1S,2S,5R,6R,7R,10S,12R,13R)-6,13-bis(hydroxymethyl)-2,6-dimethyltetracyclo[10.3.1.0¹,¹⁰.0²,⁷]hexadecane-5,13-diol
Synonyms: aphidicolin
Molecular Weight (g/mol): 338.49
Molecular Formula: C20H34O4
InChi Key: NOFOAYPPHIUXJR-APNQCZIXSA-N
SMILES: C[C@@]1(CO)[C@H](O)CC[C@@]2(C)[C@H]1CC[C@H]1C[C@@H]3C[C@]21CC[C@]3(O)CO
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Color 
  • (2)
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results