accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOH3C

CAS RN 3900-49-0

IUPAC Name: 1,6-dimethoxynaphthalene
Synonyms: 1,6-dimethoxynaphthalene
Molecular Weight (g/mol): 188.23
Molecular Formula: C12H12O2
InChi Key: RBUFUWIWCCOVOS-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(OC)=CC=CC2=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results