accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NSOON

CAS RN 39091-01-5

IUPAC Name: 4-methyl-2-(pyridin-3-yl)-1,3-thiazole-5-carboxylate
Synonyms: 4-methyl-2-(pyridin-3-yl)-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 219.24
Molecular Formula: C10H7N2O2S
InChi Key: CJLQNROYBZEUGH-UHFFFAOYSA-M
SMILES: CC1=C(SC(=N1)C1=CC=CN=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results