missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(3-pyridyl)thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15429295
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15429295 5 g 5g
15419295 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429295 Supplier Thermo Scientific Alfa Aesar Supplier No. H51723.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39091-01-5
Molecular Formula C10H7N2O2S
Molecular Weight (g/mol) 219.24
MDL Number MFCD00171753
InChI Key CJLQNROYBZEUGH-UHFFFAOYSA-M
Synonym 4-methyl-2-pyridin-3-yl-thiazole-5-carboxylic acid, 4-methyl-2-pyridin-3-yl thiazole-5-carboxylic acid, 2-3-pyridyl-4-methylthiazole-5-carboxylic acid, 4-methyl-2-3-pyridinyl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-pyridin-3-yl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-pyridin-3-yl-thiazole-5-carboxylicacid, 4-methyl-2-pyridin-3-ylthiazole-5-carboxylic acid, 4-methyl-2-3-pyridyl-1,3-thiazole-5-carboxylic acid, pubchem13985, 4-methyl-2-pyrid-3-ylthiazole-5-carboxylic acid
PubChem CID 2763489
SMILES CC1=C(SC(=N1)C1=CC=CN=C1)C([O-])=O

Specifications

Melting Point 244°C to 248°C
Quantity 5 g
IUPAC Name 4-methyl-2-pyridin-3-yl-1,3-thiazole-5-carboxylic acid
Formula Weight 220.25
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-(3-pyridyl)thiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.