accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHHC

CAS RN 3923-52-2

IUPAC Name: 1,1-diphenylprop-2-yn-1-ol
Synonyms: 1,1-diphenylprop-2-yn-1-ol
Molecular Weight (g/mol): 208.26
Molecular Formula: C15H12O
InChi Key: SMCLTAARQYTXLW-UHFFFAOYSA-N
SMILES: OC(C#C)(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results