accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOHOFFF

CAS RN 395-35-7

IUPAC Name: 2-hydroxy-2-[4-(trifluoromethyl)phenyl]acetic acid
Synonyms: hydroxy[4-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 220.15
Molecular Formula: C9H7F3O3
InChi Key: SDGXYUQKJPFLDG-UHFFFAOYNA-N
SMILES: OC(C(O)=O)C1=CC=C(C=C1)C(F)(F)F
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results