missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)mandelic acid, 98%, ACROS Organics™

Product Code. 10053640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10053640 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10053640 Supplier Thermo Scientific Acros Supplier No. 313040010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 395-35-7
CAS Min % 97.5
CAS Max % 100.0
Melting Point 130°C to 134°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H7F3O3
Linear Formula CF3C6H4CH(OH)CO2H
MDL Number MFCD00009724
Quantity 1 g
Synonym 4-trifluoromethyl mandelic acid, 2-hydroxy-2-4-trifluoromethyl phenyl acetic acid, 4-trifluoromethylmandelic acid, hydroxy 4-trifluoromethyl phenyl acetic acid, 4-trifluoromethyl phenylglycolic acid, 2-4-trifluoromethyl phenyl-2-hydroxyacetic acid, p-trifluoromethylmandelic acid
InChI Key SDGXYUQKJPFLDG-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C(C(=O)O)O)C(F)(F)F
IUPAC Name 2-hydroxy-2-[4-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol) 220.147
PubChem CID 164593
Formula Weight 220.15
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 4-(Trifluoromethyl)mandelic acid, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.