accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOHO

CAS RN 39590-81-3

IUPAC Name: [1-(hydroxymethyl)cyclopropyl]methanol
Synonyms: [1-(hydroxymethyl)cyclopropyl]methanol
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: YAINYZJQSQEGND-UHFFFAOYSA-N
SMILES: OCC1(CO)CC1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)