accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3978-80-1

(S)OHNOHHOOOCH3CH3CH3

CAS RN 3978-80-1

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-(4-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol): 281.31
Molecular Formula: C14H19NO5
InChi Key: CNBUSIJNWNXLQQ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC=C(O)C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)