missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-tyrosine, TRC

Product Code. 30348606
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30348606 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30348606

Brand: Toronto Research Chemicals TRCB66782525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-L-tyrosine
CAS 3978-80-1
Molecular Formula C14H19NO5
Quantity 25 g
Synonym N-[(1,1-Dimethylethoxy)carbonyl]-L-tyrosine, N-Carboxy-L-tyrosine N-tert-Butyl Ester, (2S)-2-[(tert-Butoxycarbonyl)amino]-3-(4-hydroxyphenyl)propanoic Acid, (2S)-2-[(tert-Butoxycarbonyl)amino]-3-(4-hydroxyphenyl)propanoic Acid, BOC-L-tyrosine, N-(tert-Butoxycarbonyl)tyrosine, N-(tert-Butyloxycarbonyl)-L-tyrosine, N-tert-Butoxycarbonyl-L-tyrosine, NSC 334307, tert-Butoxycarbonyl-L-tyrosine
InChI Formula InChI=1S/C14H19NO5/c1-14(2,3)20-13(19)15-11(12(17)18)8-9-4-6-10(16)7-5-9/h4-7,11,16H,8H2,1-3H3,(H,15,19)(H,17,18)/t11-/m0/s1
SMILES CC(C)(C)OC(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)O
IUPAC Name (2S)-3-(4-hydroxyphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Molecular Weight (g/mol) 281.3
Formula Weight 281.1263
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.