CAS RN 3983-45-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fenchlorphos-oxon, TRC
CAS: 3983-45-7 Molecular Formula: C8 H8 Cl3 O4 P Molecular Weight (g/mol): 305.48 Synonym: Phosphoric acid, dimethyl 2,4,5-trichlorophenyl ester,Dimethyl 2,4,5-trichlorophenyl phosphate,Dowco 101,Fenchlorphos-oxon,NSC 133018,O,O-Dimethyl O-2,4,5-trichlorophenyl phosphate,Ronnel oxon,Ronnel oxygen analog,Ronnoxon,Ronoxon IUPAC Name: dimethyl (2,4,5-trichlorophenyl) phosphate SMILES: COP(=O)(OC)Oc1cc(Cl)c(Cl)cc1Cl