missing translation for 'onlineSavingsMsg'
Learn More

Fenchlorphos-oxon, TRC

Product Code. 30400774
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400774 100 mg
30396130 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400774 Supplier Toronto Research Chemicals Supplier No. TRCF247410100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3983-45-7
Molecular Formula C8 H8 Cl3 O4 P
Molecular Weight (g/mol) 305.48
Synonym Phosphoric acid, dimethyl 2,4,5-trichlorophenyl ester, Dimethyl 2,4,5-trichlorophenyl phosphate, Dowco 101, Fenchlorphos-oxon, NSC 133018, O,O-Dimethyl O-2,4,5-trichlorophenyl phosphate, Ronnel oxon, Ronnel oxygen analog, Ronnoxon, Ronoxon
IUPAC Name dimethyl (2,4,5-trichlorophenyl) phosphate
SMILES COP(=O)(OC)Oc1cc(Cl)c(Cl)cc1Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H8Cl3O4P/c1-13-16(12,14-2)15-8-4-6(10)5(9)3-7(8)11/h3-4H,1-2H3
Formula Weight 303.9226
Chemical Name or Material Fenchlorphos-oxon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.