CAS RN 39969-28-3
IUPAC Name:
1-[2-(4-methoxyphenyl)ethynyl]-4-pentylbenzene
Synonyms:
1-[2-(4-methoxyphenyl)ethynyl]-4-pentylbenzene
Molecular Weight (g/mol):
278.4
Molecular Formula:
C20H22O
InChi Key:
ULPSMBQBIIZGAI-UHFFFAOYSA-N
SMILES:
CCCCCC1=CC=C(C=C1)C#CC1=CC=C(OC)C=C1
1
–
1
of
1
results
1
1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene, 99+%, Thermo Scientific™
CAS: 39969-28-3 Molecular Formula: C20H22O Molecular Weight (g/mol): 278.395 MDL Number: MFCD00187208 InChI Key: ULPSMBQBIIZGAI-UHFFFAOYSA-N Synonym: 1-2-4-methoxyphenyl ethynyl-4-pentylbenzene,1-4-methoxyphenyl ethynyl-4-n-pentylbenzene,1-4-methoxyphenyl-2-4-n-pentylphenyl-acetylene,1-methoxy-4-4-pentylphenyl ethynyl benzene,4-methoxy-1-2-4-pentylphenyl ethynyl benzene,1-methoxy-4-4-pentylphenylethynyl benzene,methyl 4-4-pentylphenyl ethynyl phenyl ether,1-methoxy-4-2-4-pentylphenyl ethynyl benzene PubChem CID: 613607 IUPAC Name: 1-methoxy-4-[2-(4-pentylphenyl)ethynyl]benzene SMILES: CCCCCC1=CC=C(C=C1)C#CC2=CC=C(C=C2)OC