missing translation for 'onlineSavingsMsg'
Learn More

1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene, 99+%, Thermo Scientific™

Product Code. 11377519
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377519 10 g 10g
11367519 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377519 Supplier Thermo Scientific Alfa Aesar Supplier No. H27889.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39969-28-3
Molecular Formula C20H22O
Molecular Weight (g/mol) 278.395
MDL Number MFCD00187208
InChI Key ULPSMBQBIIZGAI-UHFFFAOYSA-N
Synonym 1-2-4-methoxyphenyl ethynyl-4-pentylbenzene, 1-4-methoxyphenyl ethynyl-4-n-pentylbenzene, 1-4-methoxyphenyl-2-4-n-pentylphenyl-acetylene, 1-methoxy-4-4-pentylphenyl ethynyl benzene, 4-methoxy-1-2-4-pentylphenyl ethynyl benzene, 1-methoxy-4-4-pentylphenylethynyl benzene, methyl 4-4-pentylphenyl ethynyl phenyl ether, 1-methoxy-4-2-4-pentylphenyl ethynyl benzene
PubChem CID 613607
IUPAC Name 1-methoxy-4-[2-(4-pentylphenyl)ethynyl]benzene
SMILES CCCCCC1=CC=C(C=C1)C#CC2=CC=C(C=C2)OC

Specifications

Melting Point 47°C (N)
Boiling Point 168°C to 170°C (0.5 mmHg)
Quantity 10 g
Formula Weight 278.39
Percent Purity ≥99%
Chemical Name or Material 1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.