accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3997-89-5

O

CAS RN 3997-89-5

IUPAC Name: cyclopentanone
Synonyms: cyclopentanone ketopentamethylene adipic ketone adipinketon dumasin
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: BGTOWKSIORTVQH-UHFFFAOYSA-N
SMILES: O=C1CCCC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)