accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3NHHNCH3CH3

CAS RN 4013-94-9

IUPAC Name: (propan-2-yl)({2-[(propan-2-yl)amino]ethyl})amine
Synonyms: isopropyl[2-(isopropylamino)ethyl]amine
Molecular Weight (g/mol): 144.26
Molecular Formula: C8H20N2
InChi Key: MFIGJRRHGZYPDD-UHFFFAOYSA-N
SMILES: CC(C)NCCNC(C)C
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results