missing translation for 'onlineSavingsMsg'
Learn More

N,N’-Diisopropylethylenediamine, TRC

Product Code. 30180263
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180263 25 g 25g
30179850 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180263 Supplier Toronto Research Chemicals Supplier No. TRCD45528525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4013-94-9
Molecular Formula C8 H20 N2
Molecular Weight (g/mol) 144.26
Synonym N1,N2-Bis(1-methylethyl)-1,2-ethanediamine, N,N’-Bis(1-methylethyl)-1,2-ethanediamine, N,N'-Diisopropylethylenediamine, 1,2-Bis(isopropylamino)ethane, N,N’-Diisopropyl-1,2-diaminoethane, N,N’-Diisopropyl-1,2-ethanediamine, Potassium Clavulanate EP Impurity L;
IUPAC Name N,N'-di(propan-2-yl)ethane-1,2-diamine
SMILES CC(C)NCCNC(C)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H20N2/c1-7(2)9-5-6-10-8(3)4/h7-10H,5-6H2,1-4H3
Formula Weight 144.1626
Chemical Name or Material N,N'-Diisopropylethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.