accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOF

CAS RN 40273-47-0

IUPAC Name: 3-fluoropyridine-4-carbaldehyde
Synonyms: 3-fluoropyridine-4-carbaldehyde
Molecular Weight (g/mol): 125.1
Molecular Formula: C6H4FNO
InChi Key: KLKQXQXSZCNWLZ-UHFFFAOYSA-N
SMILES: FC1=C(C=O)C=CN=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results