accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ON

CAS RN 40320-60-3

IUPAC Name: 1-(diphenylmethyl)azetidin-3-one
Synonyms: 1-(diphenylmethyl)azetidin-3-one
Molecular Weight (g/mol): 237.3
Molecular Formula: C16H15NO
InChi Key: AVUDXLOVIBJFQA-UHFFFAOYSA-N
SMILES: O=C1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results