accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 405-79-8

HOOOF

CAS RN 405-79-8

IUPAC Name: 2-(4-fluorophenoxy)acetic acid
Synonyms: P-fluorophenoxyacetic acid M-fluorophenoxyacetic acid
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: ZBIULCVFFJJYTN-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results