missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Fluorophenoxyacetic acid, 98+%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Specifications
Specifications
| Melting Point | 104°C to 105°C |
| Quantity | 25 g |
| Beilstein | 2259128 |
| Formula Weight | 170.14 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 4-Fluorophenoxyacetic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction