accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2COO

CAS RN 40663-68-1

IUPAC Name: 4-(prop-2-en-1-yloxy)benzaldehyde
Synonyms: 4-(prop-2-en-1-yloxy)benzaldehyde
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: TYNJQOJWNMZQFZ-UHFFFAOYSA-N
SMILES: C=CCOC1=CC=C(C=O)C=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)