accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4076-36-2

CH3NNHNN

CAS RN 4076-36-2

IUPAC Name: 5-methyl-2H-1,2,3,4-tetrazole
Synonyms: 5-methyl-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 84.08
Molecular Formula: C2H4N4
InChi Key: XZGLNCKSNVGDNX-UHFFFAOYSA-N
SMILES: CC1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results